Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 11.94 mm Diameter, 2.0000 in OAL, Tool Steel
Catalog No. 5000512587
Encompass
This Class ZZ Plus Plug Gage is designed for precise go inspection and for setting micrometers, verniers, and other measuring devices. It features a diameter of 11.94 mm and an overall length of 2.0000 inches, crafted from durable tool steel for reliability in demanding applications. Each gage includes a certificate of accuracy with NIST traceability, ensuring compliance with ASME B89.1 standards.
  • Class ZZ Plus for precise measurement.
  • Diameter of 11.94 mm.
  • Overall length of 2.0000 inches.
  • Tolerance of 0.00508 mm.
  • Constructed from durable tool steel.
  • Includes certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Ideal for go inspection applications.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-12587 Supplier MSC Supplier No. 68102995
This Class ZZ Plus Plug Gage is designed for precise go inspection and for setting micrometers, verniers, and other measuring devices. It features a diameter of 11.94 mm and an overall length of 2.0000 inches, crafted from durable tool steel for reliability in demanding applications. Each gage includes a certificate of accuracy with NIST traceability, ensuring compliance with ASME B89.1 standards.
  • Class ZZ Plus for precise measurement.
  • Diameter of 11.94 mm.
  • Overall length of 2.0000 inches.
  • Tolerance of 0.00508 mm.
  • Constructed from durable tool steel.
  • Includes certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Ideal for go inspection applications.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Features a 10 micro coating/finish.