Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 13.53 mm, Tool Steel
Catalog No. 5000527300
Encompass
This industrial plug gage is designed for precise measurement and inspection tasks. Constructed from durable tool steel with a 10 micro finish, it ensures accuracy and reliability in various applications. Each gage includes a certificate of accuracy with NIST traceability and complies with ASME B89.1 standards.
  • Used for go inspection
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Constructed from tool steel for durability
  • Features a 10 micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
Catalog No. 50-005-27300 Supplier MSC Supplier No. 68104587
This industrial plug gage is designed for precise measurement and inspection tasks. Constructed from durable tool steel with a 10 micro finish, it ensures accuracy and reliability in various applications. Each gage includes a certificate of accuracy with NIST traceability and complies with ASME B89.1 standards.
  • Used for go inspection
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Constructed from tool steel for durability
  • Features a 10 micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards