Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 13.59 mm, 0.00508 mm Tolerance
Catalog No. 5000530975
Encompass
This Class ZZ Plus Plug Gage is designed for Go inspection and is suitable for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, it features a precise 13.59 mm diameter with a 0.00508 mm tolerance, ensuring accurate measurements. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Designed for Go inspection.
  • Used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Made from durable tool steel.
  • Features a 10 micro coating/finish.
  • Conforms to ASME B89.1 standards.
Catalog No. 50-005-30975 Supplier MSC Supplier No. 68104645
This Class ZZ Plus Plug Gage is designed for Go inspection and is suitable for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, it features a precise 13.59 mm diameter with a 0.00508 mm tolerance, ensuring accurate measurements. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Designed for Go inspection.
  • Used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Made from durable tool steel.
  • Features a 10 micro coating/finish.
  • Conforms to ASME B89.1 standards.