Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 13.87 mm Diameter, 0.00508 mm Tolerance, Tool Steel
Catalog No. 5000530995
Encompass
This plug gage is designed for precise "Go" inspection and for setting various measuring devices such as micrometers and verniers. It features a specific diameter and tolerance, manufactured from durable tool steel, ensuring accuracy and longevity in demanding laboratory and industrial applications.
  • Designed for "Go" inspection and setting measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Constructed from tool steel
  • Diameter of 13.87 mm
  • Tolerance of 0.00508 mm
  • Overall length of 2.0000 inches
Catalog No. 50-005-30995 Supplier MSC Supplier No. 68104926
This plug gage is designed for precise "Go" inspection and for setting various measuring devices such as micrometers and verniers. It features a specific diameter and tolerance, manufactured from durable tool steel, ensuring accuracy and longevity in demanding laboratory and industrial applications.
  • Designed for "Go" inspection and setting measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Constructed from tool steel
  • Diameter of 13.87 mm
  • Tolerance of 0.00508 mm
  • Overall length of 2.0000 inches