Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 13.95 mm Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000528002
Encompass
This Class ZZ Plus Plug Gage features a 13.95 mm diameter and a 2.0000 inch overall length. It offers a precise tolerance of 0.00508 mm and a 10 Micro finish. Constructed from tool steel, this gage is ideal for 'Go Inspection' applications and for calibrating micrometers, verniers, and other measuring devices. It comes with a Certificate of Accuracy with NIST Traceability and complies with ASME B89.1 standards.
  • Suitable for Go Inspection applications
  • Can be used for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Features a 13.95 mm diameter
  • Has an overall length of 2.0000 inches
  • Offers a precise tolerance of 0.00508 mm
  • Equipped with a 10 Micro finish
  • Made from durable tool steel
Catalog No. 50-005-28002 Supplier MSC Supplier No. 68105006
This Class ZZ Plus Plug Gage features a 13.95 mm diameter and a 2.0000 inch overall length. It offers a precise tolerance of 0.00508 mm and a 10 Micro finish. Constructed from tool steel, this gage is ideal for 'Go Inspection' applications and for calibrating micrometers, verniers, and other measuring devices. It comes with a Certificate of Accuracy with NIST Traceability and complies with ASME B89.1 standards.
  • Suitable for Go Inspection applications
  • Can be used for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Features a 13.95 mm diameter
  • Has an overall length of 2.0000 inches
  • Offers a precise tolerance of 0.00508 mm
  • Equipped with a 10 Micro finish
  • Made from durable tool steel