Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 14.17 mm, Tool Steel, ASME B89.1
Catalog No. 5000536366
Encompass
This Class ZZ Plus Plug Gage is designed for precise go inspection and setting measuring devices. It features a 14.17 mm diameter and an overall length of 2.0000 inches. Constructed from durable tool steel, it meets ASME B89.1 standards and offers a tight tolerance of 0.00508 mm.
  • Diameter of 14.17 mm
  • Overall length of 2.0000 inches
  • Tolerance of 0.00508 mm
  • Made from tool steel
  • 10 micro coating/finish
  • Designed for go inspection; setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
Catalog No. 50-005-36366 Supplier MSC Supplier No. 68105220
This Class ZZ Plus Plug Gage is designed for precise go inspection and setting measuring devices. It features a 14.17 mm diameter and an overall length of 2.0000 inches. Constructed from durable tool steel, it meets ASME B89.1 standards and offers a tight tolerance of 0.00508 mm.
  • Diameter of 14.17 mm
  • Overall length of 2.0000 inches
  • Tolerance of 0.00508 mm
  • Made from tool steel
  • 10 micro coating/finish
  • Designed for go inspection; setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards