Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 14.90 mm Dia, 2.0000 in OAL
Catalog No. 5000530684
Encompass
This Class ZZ Plus plug gage is designed with a 0.00508 mm tolerance and a 10 micro finish. It is primarily used for Go inspection and for setting various measuring devices such as micrometers and verniers. Each gage includes a certificate of accuracy with NIST traceability.
  • Intended for Go inspection
  • Used for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Tolerance of 0.00508 mm
  • 10 micro finish
  • Constructed from tool steel
  • Meets ASME B89.1 standards
  • Overall length of 2.0000 in
  • Diameter of measurement is 14.90 mm
Catalog No. 50-005-30684 Supplier MSC Supplier No. 68105956
This Class ZZ Plus plug gage is designed with a 0.00508 mm tolerance and a 10 micro finish. It is primarily used for Go inspection and for setting various measuring devices such as micrometers and verniers. Each gage includes a certificate of accuracy with NIST traceability.
  • Intended for Go inspection
  • Used for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Tolerance of 0.00508 mm
  • 10 micro finish
  • Constructed from tool steel
  • Meets ASME B89.1 standards
  • Overall length of 2.0000 in
  • Diameter of measurement is 14.90 mm