Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 15.58 mm Diameter, 2.00 in Overall Length, Tool Steel
Catalog No. 5000531408
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurements and inspection. With a 15.58 mm diameter and 2.00 in overall length, it is made from durable tool steel. It is suitable for Go Inspection and for setting various measuring devices like micrometers and verniers, meeting ASME B89.1 standards.
  • Includes certificate of accuracy with NIST traceability.
Catalog No. 50-005-31408 Supplier MSC Supplier No. 68106632
This Class ZZ Plus Plug Gage is designed for precise measurements and inspection. With a 15.58 mm diameter and 2.00 in overall length, it is made from durable tool steel. It is suitable for Go Inspection and for setting various measuring devices like micrometers and verniers, meeting ASME B89.1 standards.
  • Includes certificate of accuracy with NIST traceability.