Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 16.26 mm, Tool Steel
Catalog No. 5000527295
Encompass
This Class ZZ Plus plug gage is a general-purpose pin gage designed for quick and accurate measurements in various applications, including hole sizing, distance calculation between holes, slot width gaging, and Go/NoGo gaging. It features a 16.26 mm diameter, 2.0000 inch overall length, and a 0.00508 mm tolerance, ensuring reliable performance and extended wear life.
  • Economical and general-purpose pin gage
  • Used for quick and accurate measurements
  • Suitable for measuring hole sizes and distances between holes
  • Ideal for Go/NoGo gaging
  • Made from tool steel with 60/65 Rc hardness
  • Features a 10 microinch finish or better
  • Tolerance of 0.00508 mm
  • Standard length is 2 inches
  • Includes a certificate of accuracy
Catalog No. 50-005-27295 Supplier MSC Supplier No. 68107317
This Class ZZ Plus plug gage is a general-purpose pin gage designed for quick and accurate measurements in various applications, including hole sizing, distance calculation between holes, slot width gaging, and Go/NoGo gaging. It features a 16.26 mm diameter, 2.0000 inch overall length, and a 0.00508 mm tolerance, ensuring reliable performance and extended wear life.
  • Economical and general-purpose pin gage
  • Used for quick and accurate measurements
  • Suitable for measuring hole sizes and distances between holes
  • Ideal for Go/NoGo gaging
  • Made from tool steel with 60/65 Rc hardness
  • Features a 10 microinch finish or better
  • Tolerance of 0.00508 mm
  • Standard length is 2 inches
  • Includes a certificate of accuracy