Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 16.58 mm Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000531856
Encompass
This Vermont Gage Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a 16.58 mm diameter and a 2.0000 inch overall length, crafted from durable tool steel.
  • Includes certificate of accuracy with NIST traceability.
  • Used for go inspection, setting micrometers, verniers, and other measuring devices.
  • Meets ASME B89.1 standards.
  • Features a 10 micro coating/finish.
  • Constructed from tool steel.
Catalog No. 50-005-31856 Supplier MSC Supplier No. 68107630
This Vermont Gage Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a 16.58 mm diameter and a 2.0000 inch overall length, crafted from durable tool steel.
  • Includes certificate of accuracy with NIST traceability.
  • Used for go inspection, setting micrometers, verniers, and other measuring devices.
  • Meets ASME B89.1 standards.
  • Features a 10 micro coating/finish.
  • Constructed from tool steel.