Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 16.70 mm Diameter, 2.00 in Overall Length
Catalog No. 5000527680
Encompass
This precision measuring instrument is designed for accurate "Go" inspection, and for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, this plug gage features a 16.70 mm diameter with a stringent 0.00508 mm tolerance, ensuring high precision for critical applications.
  • Reliable and long-lasting due to its 2.00 in overall length and 10 Micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability for assured calibration.
  • Manufactured in the United States, meeting ASME B89.1 standards.
  • Offers a dependable solution for industrial measurement needs.
Catalog No. 50-005-27680 Supplier MSC Supplier No. 68107754
This precision measuring instrument is designed for accurate "Go" inspection, and for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, this plug gage features a 16.70 mm diameter with a stringent 0.00508 mm tolerance, ensuring high precision for critical applications.
  • Reliable and long-lasting due to its 2.00 in overall length and 10 Micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability for assured calibration.
  • Manufactured in the United States, meeting ASME B89.1 standards.
  • Offers a dependable solution for industrial measurement needs.