Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 17.43 mm Dia, Tool Steel
Catalog No. 5000527758
Encompass
This Class ZZ Plus Plug Gage features a 17.43 mm diameter and is constructed from durable tool steel with a black oxide finish. It is designed for precise 'go' inspection and can be used to set micrometers, verniers, and other measuring devices. The gage includes a certificate of accuracy with NIST traceability and conforms to ASME B89.1 standards.
  • Features a 17.43 mm diameter.
  • Constructed from durable tool steel.
  • Black oxide finish provides enhanced protection.
  • Ideal for go inspection applications.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Has an overall length of 2.0000 in.
  • Offers a tolerance of 0.00508 mm.
  • Made in the United States.
Catalog No. 50-005-27758 Supplier MSC Supplier No. 68108489
This Class ZZ Plus Plug Gage features a 17.43 mm diameter and is constructed from durable tool steel with a black oxide finish. It is designed for precise 'go' inspection and can be used to set micrometers, verniers, and other measuring devices. The gage includes a certificate of accuracy with NIST traceability and conforms to ASME B89.1 standards.
  • Features a 17.43 mm diameter.
  • Constructed from durable tool steel.
  • Black oxide finish provides enhanced protection.
  • Ideal for go inspection applications.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Has an overall length of 2.0000 in.
  • Offers a tolerance of 0.00508 mm.
  • Made in the United States.