Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 18.17 mm, 2.0000 in OAL
Catalog No. 5000536012
Encompass
The Class ZZ Plus Plug Gage is designed for precise 'Go' inspection, as well as for setting micrometers, verniers, and other measuring devices. It features an 18.17 mm diameter and a 2.0000 in overall length. Constructed from durable tool steel with a 10 micro finish, this plug gage meets ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Constructed from tool steel.
  • Features a 10 micro finish.
  • Diameter of 18.17 mm.
  • Overall length of 2.0000 in.
Catalog No. 50-005-36012 Supplier MSC Supplier No. 68109222
The Class ZZ Plus Plug Gage is designed for precise 'Go' inspection, as well as for setting micrometers, verniers, and other measuring devices. It features an 18.17 mm diameter and a 2.0000 in overall length. Constructed from durable tool steel with a 10 micro finish, this plug gage meets ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Constructed from tool steel.
  • Features a 10 micro finish.
  • Diameter of 18.17 mm.
  • Overall length of 2.0000 in.