Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 18.61 mm, 2 in
Catalog No. 5000535502
Encompass
This Class ZZ Plus plug gage has a diameter of 18.61 mm and an overall length of 2.0000 in. It features a tolerance of 0.00508 mm and a 10 Micro coating/finish. The gage is made from tool steel and is designed for "Go" inspection, as well as for setting micrometers, verniers, and other measuring devices.
  • Designed for "Go" inspection, setting micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Made from tool steel.
  • Features 10 micro coating/finish.
  • Meets ASME B89.1 standards.
Catalog No. 50-005-35502 Supplier MSC Supplier No. 68109669
This Class ZZ Plus plug gage has a diameter of 18.61 mm and an overall length of 2.0000 in. It features a tolerance of 0.00508 mm and a 10 Micro coating/finish. The gage is made from tool steel and is designed for "Go" inspection, as well as for setting micrometers, verniers, and other measuring devices.
  • Designed for "Go" inspection, setting micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Made from tool steel.
  • Features 10 micro coating/finish.
  • Meets ASME B89.1 standards.