Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 18.65 mm, Steel, Class ZZ Tolerance
Catalog No. 5000528035
Encompass
This Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement of hole sizes, ensuring accuracy in manufacturing and quality control applications. With an 18.65 mm diameter, this metric gage offers Class ZZ tolerance and is constructed from durable steel. It provides a reliable solution for verifying internal dimensions.
  • Class ZZ tolerance (.005 mm) for high precision.
  • NIST traceable for assured accuracy.
  • Made from tool steel with 60/65 Rc hardness for durability.
  • 2 inch long gage with a 10 microinch finish or better.
  • Round within .0025 mm for consistent measurements.
  • Marked with size and direction of tolerance for clear identification.
  • Certificate of Accuracy included for verification.
Catalog No. 50-005-28035 Supplier MSC Supplier No. 68109701
This Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement of hole sizes, ensuring accuracy in manufacturing and quality control applications. With an 18.65 mm diameter, this metric gage offers Class ZZ tolerance and is constructed from durable steel. It provides a reliable solution for verifying internal dimensions.
  • Class ZZ tolerance (.005 mm) for high precision.
  • NIST traceable for assured accuracy.
  • Made from tool steel with 60/65 Rc hardness for durability.
  • 2 inch long gage with a 10 microinch finish or better.
  • Round within .0025 mm for consistent measurements.
  • Marked with size and direction of tolerance for clear identification.
  • Certificate of Accuracy included for verification.