Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 19.75 mm Diameter, Tool Steel
Catalog No. 5000530687
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate inspection and calibration. Constructed from durable tool steel, it features a 19.75 mm diameter and a tight 0.00508 mm tolerance. This gage is ideal for Go inspection applications, setting micrometers, verniers, and other measuring devices, and is supplied with a Certificate of Accuracy with NIST Traceability.
  • Precisely measures 19.75 mm diameter
  • Ensures high accuracy with a 0.00508 mm tolerance
  • Constructed from durable tool steel
  • Features a 10 micro coating/finish
  • Suitable for Go inspection applications
  • Can be used to set micrometers, verniers, and other measuring devices
  • Conforms to ASME B89.1 standards
  • Includes a Certificate of Accuracy with NIST Traceability
  • Overall length of 2.0000 inches (50.8 mm)
Catalog No. 50-005-30687 Supplier MSC Supplier No. 68110808
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate inspection and calibration. Constructed from durable tool steel, it features a 19.75 mm diameter and a tight 0.00508 mm tolerance. This gage is ideal for Go inspection applications, setting micrometers, verniers, and other measuring devices, and is supplied with a Certificate of Accuracy with NIST Traceability.
  • Precisely measures 19.75 mm diameter
  • Ensures high accuracy with a 0.00508 mm tolerance
  • Constructed from durable tool steel
  • Features a 10 micro coating/finish
  • Suitable for Go inspection applications
  • Can be used to set micrometers, verniers, and other measuring devices
  • Conforms to ASME B89.1 standards
  • Includes a Certificate of Accuracy with NIST Traceability
  • Overall length of 2.0000 inches (50.8 mm)