Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 2.34 mm, 2.0000 in OAL, Tool Steel
Catalog No. 5000530358
Encompass
The Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a 2.34 mm diameter and a 2.0000 inch overall length, made from durable tool steel with a 10 Micro finish. This gage is suitable for go inspection and setting various measuring devices.
  • For go inspection; set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-30358 Supplier MSC Supplier No. 68093392
The Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a 2.34 mm diameter and a 2.0000 inch overall length, made from durable tool steel with a 10 Micro finish. This gage is suitable for go inspection and setting various measuring devices.
  • For go inspection; set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.