Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 2.67 mm Diameter, 2.00 in Overall Length
Catalog No. 5000527372
Encompass
This Class ZZ Plus Plug Gage features a 2.67 mm diameter and a 2.00 in overall length. It is designed for precise measurement applications and features a 10 Micro coating/finish, with a tolerance of 0.00508 mm.
  • For go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Constructed from tool steel
  • Complies with ASME B89.1 standards
Catalog No. 50-005-27372 Supplier MSC Supplier No. 68093723
This Class ZZ Plus Plug Gage features a 2.67 mm diameter and a 2.00 in overall length. It is designed for precise measurement applications and features a 10 Micro coating/finish, with a tolerance of 0.00508 mm.
  • For go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Constructed from tool steel
  • Complies with ASME B89.1 standards