Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 3.97 mm, 2.0000 in OAL
Catalog No. 5000527241
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement applications, including Go Inspection and setting measuring devices like micrometers and verniers. It features a 3.97 mm diameter, a 2.0000 inch overall length, and is constructed from durable tool steel. This gage ensures accuracy with a 0.00508 mm tolerance and a 10 microfinish. Each gage includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards.
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Features a 10 microfinish.
  • Constructed from durable tool steel.
  • Provides high precision with a 0.00508 mm tolerance.
Catalog No. 50-005-27241 Supplier MSC Supplier No. 68095025
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement applications, including Go Inspection and setting measuring devices like micrometers and verniers. It features a 3.97 mm diameter, a 2.0000 inch overall length, and is constructed from durable tool steel. This gage ensures accuracy with a 0.00508 mm tolerance and a 10 microfinish. Each gage includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards.
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Features a 10 microfinish.
  • Constructed from durable tool steel.
  • Provides high precision with a 0.00508 mm tolerance.