Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 4.01 mm Diameter, 2 in Overall Length, Class ZZ Tolerance
Catalog No. 5000530952
Encompass
This economical Class ZZ Plus Plug Gage is designed for quick and accurate measurements in various gaging applications. Made from durable tool steel with a 10 microinch finish, it measures hole sizes, calculates distances between holes, gages slot widths, and checks locations. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Economical general-purpose pin gage.
  • Used for quick and accurate measurements including hole sizes, distances, slot widths, and locations.
  • Suitable for go/nogo gaging, micrometers, and snap gages.
  • Offers extended wear life.
  • Includes certificate of accuracy and is NIST traceable.
  • Constructed from tool steel (60/65 Rc) with a 10 microinch finish.
  • Measures 2 in long and is round within 0.0001 in (0.0025 mm).
  • Marked with size and tolerance direction on gages over 0.0605 in.
Catalog No. 50-005-30952 Supplier MSC Supplier No. 68095066
This economical Class ZZ Plus Plug Gage is designed for quick and accurate measurements in various gaging applications. Made from durable tool steel with a 10 microinch finish, it measures hole sizes, calculates distances between holes, gages slot widths, and checks locations. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Economical general-purpose pin gage.
  • Used for quick and accurate measurements including hole sizes, distances, slot widths, and locations.
  • Suitable for go/nogo gaging, micrometers, and snap gages.
  • Offers extended wear life.
  • Includes certificate of accuracy and is NIST traceable.
  • Constructed from tool steel (60/65 Rc) with a 10 microinch finish.
  • Measures 2 in long and is round within 0.0001 in (0.0025 mm).
  • Marked with size and tolerance direction on gages over 0.0605 in.