Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 4.02 mm, 2.0000 in OAL
Catalog No. 5000531342
Encompass
This plug gage is an essential tool for precise dimensional verification and calibration. Designed for "Go" inspection, it can also be used to accurately set and calibrate micrometers, verniers, and a variety of other measuring instruments. Crafted from durable tool steel, it offers reliability and accuracy for critical measurement tasks.
  • Go inspection for quality control
  • Sets micrometers, verniers, and other measuring devices
  • Constructed from durable tool steel
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Features a 10 micro coating/finish
  • Precise 4.02 mm diameter
  • Overall length of 2.0000 in
  • Tolerance of 0.00508 mm for high accuracy
Catalog No. 50-005-31342 Supplier MSC Supplier No. 68095074
This plug gage is an essential tool for precise dimensional verification and calibration. Designed for "Go" inspection, it can also be used to accurately set and calibrate micrometers, verniers, and a variety of other measuring instruments. Crafted from durable tool steel, it offers reliability and accuracy for critical measurement tasks.
  • Go inspection for quality control
  • Sets micrometers, verniers, and other measuring devices
  • Constructed from durable tool steel
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Features a 10 micro coating/finish
  • Precise 4.02 mm diameter
  • Overall length of 2.0000 in
  • Tolerance of 0.00508 mm for high accuracy