Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 4.33 mm, 2 inch, 0.00508 mm Tolerance
Catalog No. 5000527959
Encompass
Vermont Gage Class ZZ Plus Plug Gages are economical, general-purpose pin gages used for quick and accurate measurements. They are suitable for checking hole sizes, distances between holes, slot widths, and for Go/NoGo gaging and sizing in micrometers and snap gages. These gages are designed for extended wear life.
  • NIST traceable.
  • Certificate of accuracy included.
  • Class ZZ tolerance: 0.00508 mm.
  • Plus (Go) tolerance direction.
  • Tool steel construction with 60/65 Rc hardness.
  • 2 inch length.
  • 10 microinch finish or better.
  • Round within 0.0025 mm.
  • Marked with size and direction of tolerance on gages over 0.0605 "
Catalog No. 50-005-27959 Supplier MSC Supplier No. 68095389
Vermont Gage Class ZZ Plus Plug Gages are economical, general-purpose pin gages used for quick and accurate measurements. They are suitable for checking hole sizes, distances between holes, slot widths, and for Go/NoGo gaging and sizing in micrometers and snap gages. These gages are designed for extended wear life.
  • NIST traceable.
  • Certificate of accuracy included.
  • Class ZZ tolerance: 0.00508 mm.
  • Plus (Go) tolerance direction.
  • Tool steel construction with 60/65 Rc hardness.
  • 2 inch length.
  • 10 microinch finish or better.
  • Round within 0.0025 mm.
  • Marked with size and direction of tolerance on gages over 0.0605 "