Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 4.35 mm Diameter, 0.00508 mm Tolerance
Catalog No. 5000510316
Encompass
This Class ZZ Plus Plug Gage is a high-precision measuring tool engineered for accuracy in industrial and metrology applications. With a specific diameter of 4.35 mm and a 2.0000-inch overall length, this gage is ideal for verifying bore sizes and setting other measuring instruments.
  • Features a tight tolerance of 0.00508 mm and a 10 micro finish for high accuracy.
  • Made from robust tool steel to ensure longevity and consistent performance.
  • Suitable for "go" inspection processes and for calibrating various measuring devices such as micrometers and verniers.
  • Comes with a certificate of accuracy that includes NIST traceability, affirming its adherence to rigorous quality standards.
  • Manufactured to meet ASME B89.1 standards.
  • Proudly made in the United States.
Catalog No. 50-005-10316 Supplier MSC Supplier No. 68095405
This Class ZZ Plus Plug Gage is a high-precision measuring tool engineered for accuracy in industrial and metrology applications. With a specific diameter of 4.35 mm and a 2.0000-inch overall length, this gage is ideal for verifying bore sizes and setting other measuring instruments.
  • Features a tight tolerance of 0.00508 mm and a 10 micro finish for high accuracy.
  • Made from robust tool steel to ensure longevity and consistent performance.
  • Suitable for "go" inspection processes and for calibrating various measuring devices such as micrometers and verniers.
  • Comes with a certificate of accuracy that includes NIST traceability, affirming its adherence to rigorous quality standards.
  • Manufactured to meet ASME B89.1 standards.
  • Proudly made in the United States.