Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 6.48 mm, 50.8 mm OAL, 0.00508 mm Tolerance
Catalog No. 5000511137
Encompass
Vermont Gage Class ZZ Plus Plug Gages are precision measuring tools designed for accurately checking hole sizes. This plug gage features a 6.48 mm diameter and a 50.8 mm (2.0000 in) overall length, ensuring precise measurements for quality control and inspection applications.
  • Precision ground to a 6.48 mm diameter.
  • Overall length of 50.8 mm (2.0000 in).
  • Achieves a high tolerance of 0.00508 mm for accuracy.
  • Smooth 10 microfinish for reliable measurements.
  • Class ZZ Plus classification meets stringent quality requirements.
Catalog No. 50-005-11137 Supplier MSC Supplier No. 68097534
Vermont Gage Class ZZ Plus Plug Gages are precision measuring tools designed for accurately checking hole sizes. This plug gage features a 6.48 mm diameter and a 50.8 mm (2.0000 in) overall length, ensuring precise measurements for quality control and inspection applications.
  • Precision ground to a 6.48 mm diameter.
  • Overall length of 50.8 mm (2.0000 in).
  • Achieves a high tolerance of 0.00508 mm for accuracy.
  • Smooth 10 microfinish for reliable measurements.
  • Class ZZ Plus classification meets stringent quality requirements.