Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 6.68 mm, Tool Steel
Catalog No. 5000531890
Encompass
This product is a Class ZZ Plus Plug Gage, with a diameter of 6.68 mm and an overall length of 2.0000 in. It features a tolerance of 0.00508 mm and a 10 Micro finish. The gage is made from tool steel and is designed for "Go" inspection, as well as for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Made from durable tool steel material.
  • Suitable for "Go" inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
Catalog No. 50-005-31890 Supplier MSC Supplier No. 68097732
This product is a Class ZZ Plus Plug Gage, with a diameter of 6.68 mm and an overall length of 2.0000 in. It features a tolerance of 0.00508 mm and a 10 Micro finish. The gage is made from tool steel and is designed for "Go" inspection, as well as for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Made from durable tool steel material.
  • Suitable for "Go" inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.