Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 6.85 mm, 2.0000 in
Catalog No. 5000531401
Encompass
This Vermont Gage Class ZZ Plus Plug Gage is designed for precision 'go' inspection and for setting measuring devices such as micrometers and verniers. Constructed from durable tool steel, it features a 6.85 mm diameter and a 2.0000 inch overall length.
  • For go inspection and setting measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • 0.00508 mm tolerance
  • 10 micro finish
  • Meets ASME B89.1 standards
  • Made in the United States
Catalog No. 50-005-31401 Supplier MSC Supplier No. 68097906
This Vermont Gage Class ZZ Plus Plug Gage is designed for precision 'go' inspection and for setting measuring devices such as micrometers and verniers. Constructed from durable tool steel, it features a 6.85 mm diameter and a 2.0000 inch overall length.
  • For go inspection and setting measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • 0.00508 mm tolerance
  • 10 micro finish
  • Meets ASME B89.1 standards
  • Made in the United States