Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 7.30 mm Dia, 2.0000 in OAL
Catalog No. 5000528928
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise go inspection and for setting measuring devices such as micrometers and verniers. It features a 7.30 mm diameter and a 2.0000 inch overall length.
  • Tool steel construction.
  • 7.30 mm diameter.
  • 2.0000 inch overall length.
  • Tolerance of 0.00508 mm.
  • 10 micro finish.
  • Class ZZ plus accuracy.
  • Manufactured to ASME B89.1 standards.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices.
Catalog No. 50-005-28928 Supplier MSC Supplier No. 68098359
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise go inspection and for setting measuring devices such as micrometers and verniers. It features a 7.30 mm diameter and a 2.0000 inch overall length.
  • Tool steel construction.
  • 7.30 mm diameter.
  • 2.0000 inch overall length.
  • Tolerance of 0.00508 mm.
  • 10 micro finish.
  • Class ZZ plus accuracy.
  • Manufactured to ASME B89.1 standards.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices.