Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 8.00 mm, 2.00 in OAL, Tool Steel, 0.00508 mm Tolerance
Catalog No. 5000530584
Encompass
This Class ZZ Plus Plug Gage features an 8.00 mm diameter and a 2.00 in overall length. Manufactured from tool steel, it offers a 0.00508 mm tolerance and a 10 Micro finish. It is designed for Go Inspection and calibrating various measuring devices, adhering to ASME B89.1 standards.
  • Includes Certificate of Accuracy with NIST traceability.
  • Designed for go inspection, setting micrometers, verniers, and other measuring devices.
  • Constructed from tool steel for durability.
  • Features a 10 micro finish.
  • Adheres to ASME B89.1 standards.
Catalog No. 50-005-30584 Supplier MSC Supplier No. 68099050
This Class ZZ Plus Plug Gage features an 8.00 mm diameter and a 2.00 in overall length. Manufactured from tool steel, it offers a 0.00508 mm tolerance and a 10 Micro finish. It is designed for Go Inspection and calibrating various measuring devices, adhering to ASME B89.1 standards.
  • Includes Certificate of Accuracy with NIST traceability.
  • Designed for go inspection, setting micrometers, verniers, and other measuring devices.
  • Constructed from tool steel for durability.
  • Features a 10 micro finish.
  • Adheres to ASME B89.1 standards.