Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 8.47 mm Diameter, 0.00508 mm Tolerance
Catalog No. 5000530667
Encompass
This Class ZZ Plus Plug Gage from Vermont Gage features a diameter of 8.47 mm and a tolerance of 0.00508 mm. Constructed from tool steel, it is designed for 'Go' inspection, setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability and meets ASME B89.1 standards.
  • Used for Go inspection
  • Set micrometers
  • Set verniers and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Constructed from tool steel
  • Features a 10 Micro finish
Catalog No. 50-005-30667 Supplier MSC Supplier No. 68099522
This Class ZZ Plus Plug Gage from Vermont Gage features a diameter of 8.47 mm and a tolerance of 0.00508 mm. Constructed from tool steel, it is designed for 'Go' inspection, setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability and meets ASME B89.1 standards.
  • Used for Go inspection
  • Set micrometers
  • Set verniers and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Constructed from tool steel
  • Features a 10 Micro finish