Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 8.75 mm, 2.0000 in
Catalog No. 5000530956
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Made from durable tool steel, it offers accurate gaging for various applications, including setting micrometers and verniers.
  • For Go inspection; set micrometers; verniers & other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Tool steel material
  • 8.75 mm diameter
  • 2.0000 in overall length
  • 0.00508 mm tolerance
  • 10 micro coating/finish
  • Country of origin: United States
Catalog No. 50-005-30956 Supplier MSC Supplier No. 68099803
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Made from durable tool steel, it offers accurate gaging for various applications, including setting micrometers and verniers.
  • For Go inspection; set micrometers; verniers & other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Tool steel material
  • 8.75 mm diameter
  • 2.0000 in overall length
  • 0.00508 mm tolerance
  • 10 micro coating/finish
  • Country of origin: United States