Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 8.80 mm, 0.00508 mm Tolerance
Catalog No. 5000537277
Encompass
Class ZZ Plus Plug Gages are precise measuring tools used for 'go' inspection and for accurately setting micrometers, verniers, and other measuring devices. These gages are crafted from durable tool steel and feature a specific diameter and tight tolerance for reliable performance and accuracy.
  • Used for 'go' inspection and setting of measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Constructed from tool steel
  • Features an 8.80 mm diameter
  • Maintains a 0.00508 mm tolerance
  • Finished with a 10 microfinish
  • Has an overall length of 2.00 in
Catalog No. 50-005-37277 Supplier MSC Supplier No. 68099852
Class ZZ Plus Plug Gages are precise measuring tools used for 'go' inspection and for accurately setting micrometers, verniers, and other measuring devices. These gages are crafted from durable tool steel and feature a specific diameter and tight tolerance for reliable performance and accuracy.
  • Used for 'go' inspection and setting of measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Constructed from tool steel
  • Features an 8.80 mm diameter
  • Maintains a 0.00508 mm tolerance
  • Finished with a 10 microfinish
  • Has an overall length of 2.00 in