Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 8.85 mm, 2.0000 in OAL
Catalog No. 5000527915
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features an 8.85 mm diameter and a 2.0000 in overall length, constructed from durable tool steel. This gage is suitable for go inspection and for setting various measuring devices like micrometers and verniers.
  • Features an 8.85 mm measurement diameter.
  • Has an overall length of 2.0000 in.
  • Constructed from tool steel for durability.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Has a 10 micro coating/finish.
  • Suitable for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
Catalog No. 50-005-27915 Supplier MSC Supplier No. 68099902
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features an 8.85 mm diameter and a 2.0000 in overall length, constructed from durable tool steel. This gage is suitable for go inspection and for setting various measuring devices like micrometers and verniers.
  • Features an 8.85 mm measurement diameter.
  • Has an overall length of 2.0000 in.
  • Constructed from tool steel for durability.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Has a 10 micro coating/finish.
  • Suitable for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.