Learn More
MSC Vermont Gage Gage, Class ZZ Minus, 2.26 mm, 2 in
Catalog No. 5000534044
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications such as setting micrometers, no-go inspection, and calibrating verniers and other measuring devices. Constructed from steel, it features a 2.26 mm diameter and a 2 inch overall length, ensuring accuracy and durability for various laboratory and industrial tasks.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from steel for durability.
  • 10 micro coating/finish.
  • Suitable for no-go inspection.
  • Ideal for setting micrometers and calibrating measuring devices.
Catalog No. 50-005-34044 Supplier MSC Supplier No. 40429714
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications such as setting micrometers, no-go inspection, and calibrating verniers and other measuring devices. Constructed from steel, it features a 2.26 mm diameter and a 2 inch overall length, ensuring accuracy and durability for various laboratory and industrial tasks.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from steel for durability.
  • 10 micro coating/finish.
  • Suitable for no-go inspection.
  • Ideal for setting micrometers and calibrating measuring devices.