Learn More
MSC Vermont Gage Gage Pins & Sets, Class ZZ, Tool Steel, 2 in Long
Catalog No. 5000512544
Encompass
Vermont Gage Steel Class ZZ Gage Pins & Sets are designed for precise measurement and inspection applications. Crafted from durable tool steel, these 2-inch long pins provide accurate results for various gaging tasks, ensuring quality control in manufacturing and scientific environments.
  • Make quick and accurate measurements
  • Measure hole diameters, calculate distances between holes, and gage slot widths and locations
  • Use for Go/NoGo inspection
  • Set micrometers, verniers, and other measuring devices
  • Certificate of Accuracy is included
  • ISO 17025 accredited and non-accredited calibration certificates may be purchased
Catalog No. 50-005-12544 Supplier MSC Supplier No. 62255609
Vermont Gage Steel Class ZZ Gage Pins & Sets are designed for precise measurement and inspection applications. Crafted from durable tool steel, these 2-inch long pins provide accurate results for various gaging tasks, ensuring quality control in manufacturing and scientific environments.
  • Make quick and accurate measurements
  • Measure hole diameters, calculate distances between holes, and gage slot widths and locations
  • Use for Go/NoGo inspection
  • Set micrometers, verniers, and other measuring devices
  • Certificate of Accuracy is included
  • ISO 17025 accredited and non-accredited calibration certificates may be purchased