Learn More
MSC Vermont Gage Go Plug Gage, 0.1572 in., Class X, 2 in. Length
Catalog No. 5000534082
Encompass
This Go Plug Gage features a 0.1572 in. diameter and Class X tolerance, suitable for precision measurement applications. It is crafted from cold stabilized Tool Steel and comes with NIST traceability, ensuring high accuracy for critical quality control tasks.
  • Class X NIST traceable
  • Made from Tool Steel; 60/65 Rc (Cold Stabilized)
  • 2 in. long
  • 4 microinch finish or better
  • Round within 1/2 of tolerance
  • Includes Certificate of Accuracy
Catalog No. 50-005-34082 Supplier MSC Supplier No. 01114537
This Go Plug Gage features a 0.1572 in. diameter and Class X tolerance, suitable for precision measurement applications. It is crafted from cold stabilized Tool Steel and comes with NIST traceability, ensuring high accuracy for critical quality control tasks.
  • Class X NIST traceable
  • Made from Tool Steel; 60/65 Rc (Cold Stabilized)
  • 2 in. long
  • 4 microinch finish or better
  • Round within 1/2 of tolerance
  • Includes Certificate of Accuracy