Learn More
MSC Vermont Gage Metric Pin Gage, Class ZZ, 2 in, Tool steel, .02 mm Increments
Catalog No. 5000514432
Encompass
Vermont Gage Standard Class ZZ Pin Gages provide quick and accurate measurements for hole sizes, distances, and slot widths. These economical gages are ideal for Go/NoGo gaging and offer extended wear life, making them suitable for various general-purpose applications.
  • Economical and versatile for various gaging applications.
  • Provides quick and precise measurements of hole sizes, distances, and slot widths.
  • Facilitates Go/NoGo inspection for determining part acceptance.
  • Features extended wear life for enhanced longevity.
  • NIST traceable, ensuring accuracy and compliance.
  • Available in durable Tool Steel or Black Guard™ materials with a high-quality 10 microinch finish.
  • Includes a Certificate of Accuracy.
  • Calibration services are offered for sets and libraries.
Catalog No. 50-005-14432 Supplier MSC Supplier No. 40439143
Vermont Gage Standard Class ZZ Pin Gages provide quick and accurate measurements for hole sizes, distances, and slot widths. These economical gages are ideal for Go/NoGo gaging and offer extended wear life, making them suitable for various general-purpose applications.
  • Economical and versatile for various gaging applications.
  • Provides quick and precise measurements of hole sizes, distances, and slot widths.
  • Facilitates Go/NoGo inspection for determining part acceptance.
  • Features extended wear life for enhanced longevity.
  • NIST traceable, ensuring accuracy and compliance.
  • Available in durable Tool Steel or Black Guard™ materials with a high-quality 10 microinch finish.
  • Includes a Certificate of Accuracy.
  • Calibration services are offered for sets and libraries.