Learn More
MSC Vermont Gage Metric Pin Gage Set, 1.30 to 4.98 mm, Class ZZ, 0.005 mm Tolerance
Catalog No. 5000511866
Encompass
Vermont Gage Metric Pin Gages are precision tools utilized for accurately measuring bore and hole diameters. These sets are meticulously designed with specific increments and tolerances to ensure high precision in a wide array of industrial and manufacturing applications. Featuring a Class ZZ tolerance and a 0.005 mm deviation, these pin gages guarantee reliable measurements essential for rigorous quality control and inspection processes.
  • Metric sizes for international compatibility
  • Provided in convenient sets covering a broad range of measurements
  • Class ZZ tolerance for high-precision applications
  • Constructed from high-quality material for enhanced durability
  • Ensures accurate verification of bore and hole diameters
Catalog No. 50-005-11866 Supplier MSC Supplier No. 40446296
Vermont Gage Metric Pin Gages are precision tools utilized for accurately measuring bore and hole diameters. These sets are meticulously designed with specific increments and tolerances to ensure high precision in a wide array of industrial and manufacturing applications. Featuring a Class ZZ tolerance and a 0.005 mm deviation, these pin gages guarantee reliable measurements essential for rigorous quality control and inspection processes.
  • Metric sizes for international compatibility
  • Provided in convenient sets covering a broad range of measurements
  • Class ZZ tolerance for high-precision applications
  • Constructed from high-quality material for enhanced durability
  • Ensures accurate verification of bore and hole diameters