Learn More
MSC Vermont Gage Metric Pin Gage Set, Class ZZ, 0.005 mm Tolerance
Catalog No. 5000516499
Encompass
Vermont Gage Metric Pin Gage Sets are precision tools used for accurate measurement in various applications. These sets are available in a wide range of metric sizes and are designed with a Class ZZ tolerance of 0.005 mm, ensuring high accuracy for critical gaging tasks.
  • Available in various metric ranges to suit diverse gaging needs.
  • Each set features a Class ZZ tolerance of 0.005 mm for high precision.
  • Includes different numbers of pins per set depending on the size range.
  • Offered in Plus and Minus variations.
  • Available in steel and black guard models.
Catalog No. 50-005-16499 Supplier MSC Supplier No. 40431322
Vermont Gage Metric Pin Gage Sets are precision tools used for accurate measurement in various applications. These sets are available in a wide range of metric sizes and are designed with a Class ZZ tolerance of 0.005 mm, ensuring high accuracy for critical gaging tasks.
  • Available in various metric ranges to suit diverse gaging needs.
  • Each set features a Class ZZ tolerance of 0.005 mm for high precision.
  • Includes different numbers of pins per set depending on the size range.
  • Offered in Plus and Minus variations.
  • Available in steel and black guard models.