Learn More
MSC Vermont Gage Metric Pin Gage Set, Class ZZ, Steel, 0.20mm to 1.28mm
Catalog No. 5000514331
Encompass
These pin gage sets are economical, general-purpose tools designed for quick and accurate measurements in various applications. They are suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations, as well as Go/NoGo gaging and sizing in micrometers and snap gages. Constructed from tool steel, these gages offer added wear life.
  • Designed for quick and accurate measurements
  • Suitable for measuring hole sizes and distances between holes
  • Ideal for gaging slot widths and checking locations
  • Usable for Go/NoGo gaging and sizing in micrometers and snap gages
  • Constructed from tool steel for added wear life
  • Features a 10 microinch or better finish
  • Maintains roundness within 0.0025mm
  • Includes 55 gages in a range from 0.20mm to 1.28mm
Catalog No. 50-005-14331 Supplier MSC Supplier No. 68105386
These pin gage sets are economical, general-purpose tools designed for quick and accurate measurements in various applications. They are suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations, as well as Go/NoGo gaging and sizing in micrometers and snap gages. Constructed from tool steel, these gages offer added wear life.
  • Designed for quick and accurate measurements
  • Suitable for measuring hole sizes and distances between holes
  • Ideal for gaging slot widths and checking locations
  • Usable for Go/NoGo gaging and sizing in micrometers and snap gages
  • Constructed from tool steel for added wear life
  • Features a 10 microinch or better finish
  • Maintains roundness within 0.0025mm
  • Includes 55 gages in a range from 0.20mm to 1.28mm