Learn More
MSC Vermont Gage No-Go Thread Ring Gage, 5/8-11 Thread, Class 2A
Catalog No. 5000530413
Encompass
This thread ring gage is engineered for precise measurement of external threads. It is designed to verify the 'No-Go' limit of 5/8-11 NC Class 2A threads, ensuring that the maximum material limit is not exceeded.
  • Verifies maximum material limits for external threads
  • Suitable for 5/8-11 thread sizes
  • Conforms to Class 2A thread standards
Catalog No. 50-005-30413 Supplier MSC Supplier No. 55323331
This thread ring gage is engineered for precise measurement of external threads. It is designed to verify the 'No-Go' limit of 5/8-11 NC Class 2A threads, ensuring that the maximum material limit is not exceeded.
  • Verifies maximum material limits for external threads
  • Suitable for 5/8-11 thread sizes
  • Conforms to Class 2A thread standards