Learn More
MSC Vermont Gage Pin Gage, 0.0220 in Diameter, Class ZZ, Tool Steel
Catalog No. 5000531538
Encompass
This Vermont Gage Class ZZ pin gage features a 0.0220 inch diameter and is constructed from durable tool steel. Designed for precise measurements, it is ideal for applications such as measuring hole diameters, gaging slot widths, and setting various measuring devices like micrometers and verniers. It also supports Go/NoGo inspection.
  • 0.0220 inch diameter
  • 2 inch overall length
  • Class ZZ tolerance of 0.0002 inches
  • Made from tool steel with 60/65 Rc hardness
  • 10 microinch finish or better
  • Roundness within 0.0001 inches
  • Suitable for measuring hole diameters and slot widths
  • Can be used for Go/NoGo inspection
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
Catalog No. 50-005-31538 Supplier MSC Supplier No. 88902200
This Vermont Gage Class ZZ pin gage features a 0.0220 inch diameter and is constructed from durable tool steel. Designed for precise measurements, it is ideal for applications such as measuring hole diameters, gaging slot widths, and setting various measuring devices like micrometers and verniers. It also supports Go/NoGo inspection.
  • 0.0220 inch diameter
  • 2 inch overall length
  • Class ZZ tolerance of 0.0002 inches
  • Made from tool steel with 60/65 Rc hardness
  • 10 microinch finish or better
  • Roundness within 0.0001 inches
  • Suitable for measuring hole diameters and slot widths
  • Can be used for Go/NoGo inspection
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards