Learn More
MSC Vermont Gage Pin Gage, 0.0320 in Diameter, 2.0000 in Length, Class ZZ, Plus, Black Guard
Catalog No. 5000511900
Encompass
This Vermont Gage pin gage features a 0.0320 in diameter and a 2.0000 in overall length. It is a Class ZZ 'Plus' (Go) gage, treated with Black Guard black oxide for enhanced durability and corrosion resistance.
  • Black Guard black oxide treatment prevents rust and corrosion, extending the useful life.
  • Visible wear indication allows for effective calibration or replacement.
  • Class ZZ tolerance of 0.0002 in (0.005 mm).
  • Constructed from tool steel with a hardness of 60 to 65 Rc.
  • Overall length of 2 in.
  • Features a 10 microinch finish or better.
  • Round within 0.0001 in (0.0025 mm).
  • Certificate of Accuracy is included.
Catalog No. 50-005-11900 Supplier MSC Supplier No. 50734227
This Vermont Gage pin gage features a 0.0320 in diameter and a 2.0000 in overall length. It is a Class ZZ 'Plus' (Go) gage, treated with Black Guard black oxide for enhanced durability and corrosion resistance.
  • Black Guard black oxide treatment prevents rust and corrosion, extending the useful life.
  • Visible wear indication allows for effective calibration or replacement.
  • Class ZZ tolerance of 0.0002 in (0.005 mm).
  • Constructed from tool steel with a hardness of 60 to 65 Rc.
  • Overall length of 2 in.
  • Features a 10 microinch finish or better.
  • Round within 0.0001 in (0.0025 mm).
  • Certificate of Accuracy is included.