Learn More
MSC Vermont Gage Pin Gage, 0.0405 in Dia, 2 in Long, Class X Minus
Catalog No. 5000510516
Encompass
This Vermont Gage Class X Minus Pin Gage is a precision tool designed for accurate measurement. Made from durable tool steel, it features a 0.0405 inch diameter and a 2 inch length. It is ideal for the calibration of measuring equipment or for direct gaging of parts, ensuring high precision in various applications.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 4 micro coated finish.
  • Suitable for calibration of measuring equipment or direct gaging.
  • Constructed from durable tool steel.
Catalog No. 50-005-10516 Supplier MSC Supplier No. 88804059
This Vermont Gage Class X Minus Pin Gage is a precision tool designed for accurate measurement. Made from durable tool steel, it features a 0.0405 inch diameter and a 2 inch length. It is ideal for the calibration of measuring equipment or for direct gaging of parts, ensuring high precision in various applications.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 4 micro coated finish.
  • Suitable for calibration of measuring equipment or direct gaging.
  • Constructed from durable tool steel.