Learn More
MSC Vermont Gage Pin Gage, 0.0410 in, Class ZZ, Minus Tolerance, Steel
Catalog No. 5000530508
Encompass
This Vermont Gage pin gage is designed for precise measurement of hole sizes. It features a 0.0410 in diameter and a Class ZZ minus tolerance of +0.0000/-0.0002 in. Constructed from steel, this gage is suitable for demanding industrial and laboratory applications. It meets ISO 17025 standards and comes with a Certificate of Accuracy with NIST Traceability.
  • Constructed from steel
  • Hardness of RC 60/65
  • Overall length of 2 in
  • Tolerance of +0.0000/-0.0002 in
  • Meets ISO 17025 standards
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-30508 Supplier MSC Supplier No. 89004105
This Vermont Gage pin gage is designed for precise measurement of hole sizes. It features a 0.0410 in diameter and a Class ZZ minus tolerance of +0.0000/-0.0002 in. Constructed from steel, this gage is suitable for demanding industrial and laboratory applications. It meets ISO 17025 standards and comes with a Certificate of Accuracy with NIST Traceability.
  • Constructed from steel
  • Hardness of RC 60/65
  • Overall length of 2 in
  • Tolerance of +0.0000/-0.0002 in
  • Meets ISO 17025 standards
  • Includes certificate of accuracy with NIST traceability