Learn More
MSC Vermont Gage Pin Gage, 0.0640 inch Diameter, 2 inch Length, Tool Steel
Catalog No. 5000514382
Encompass
This Class X Plus Pin Gage is precision-engineered for accuracy in measurement. Crafted from durable tool steel, it features a 0.0640 inch diameter and a 2 inch length. The gage is ideal for calibrating measuring equipment or directly gaging parts, ensuring reliability in critical applications.
  • Constructed from tool steel for durability.
  • Features a 4 micro-coated finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for calibration of measuring equipment or direct part gaging.
  • Maintains a tight tolerance of 0.00004 "
Catalog No. 50-005-14382 Supplier MSC Supplier No. 88706403
This Class X Plus Pin Gage is precision-engineered for accuracy in measurement. Crafted from durable tool steel, it features a 0.0640 inch diameter and a 2 inch length. The gage is ideal for calibrating measuring equipment or directly gaging parts, ensuring reliability in critical applications.
  • Constructed from tool steel for durability.
  • Features a 4 micro-coated finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for calibration of measuring equipment or direct part gaging.
  • Maintains a tight tolerance of 0.00004 "