Learn More
MSC Vermont Gage Pin Gage, 0.0715 inch, Class ZZ, Minus
Catalog No. 5000535150
Encompass
This precision pin gage provides accurate measurement for various applications. With a 0.0715 inch diameter and manufactured to Class ZZ tolerances, it ensures reliable performance for inspection and calibration tasks. This minus-tolerance pin gage is ideal for go/no-go gaging and checking bore sizes.
  • Accurate diameter for precise measurement
  • Manufactured to Class ZZ tolerances
  • Minus tolerance for specific gaging requirements
  • Suitable for inspection and calibration
  • Used for go/no-go gaging
  • Helps in checking bore sizes
Catalog No. 50-005-35150 Supplier MSC Supplier No. 50754837
This precision pin gage provides accurate measurement for various applications. With a 0.0715 inch diameter and manufactured to Class ZZ tolerances, it ensures reliable performance for inspection and calibration tasks. This minus-tolerance pin gage is ideal for go/no-go gaging and checking bore sizes.
  • Accurate diameter for precise measurement
  • Manufactured to Class ZZ tolerances
  • Minus tolerance for specific gaging requirements
  • Suitable for inspection and calibration
  • Used for go/no-go gaging
  • Helps in checking bore sizes