Learn More
MSC Vermont Gage Pin Gage, 0.0815 inch, Class ZZ Minus, 52100 Tool Steel
Catalog No. 5000533495
Encompass
This pin gage is engineered for high-accuracy measurement and inspection applications. It provides precise dimensional verification with a specific diameter and tolerance classification, essential for quality control and manufacturing processes.
  • Precisely measures small internal diameters.
  • Class ZZ Minus tolerance ensures high accuracy.
  • Constructed from durable 52100 tool steel.
  • Ideal for quality control and calibration tasks.
Catalog No. 50-005-33495 Supplier MSC Supplier No. 01587252
This pin gage is engineered for high-accuracy measurement and inspection applications. It provides precise dimensional verification with a specific diameter and tolerance classification, essential for quality control and manufacturing processes.
  • Precisely measures small internal diameters.
  • Class ZZ Minus tolerance ensures high accuracy.
  • Constructed from durable 52100 tool steel.
  • Ideal for quality control and calibration tasks.