Learn More
MSC Vermont Gage Pin Gage, 0.1010 in Dia, 2 in Long, Class X Minus
Catalog No. 5000531148
Encompass
The Vermont Gage Class X Minus Pin Gage is designed for the calibration of measuring equipment or for direct gaging of parts. This precision gage ensures accurate measurements for critical applications.
  • Includes certificate of accuracy with NIST traceability
  • Ideal for calibration of measuring equipment or for direct gaging of parts
  • Constructed from durable tool steel
  • Features a 4 micro coating/finish for enhanced performance
Catalog No. 50-005-31148 Supplier MSC Supplier No. 88810106
The Vermont Gage Class X Minus Pin Gage is designed for the calibration of measuring equipment or for direct gaging of parts. This precision gage ensures accurate measurements for critical applications.
  • Includes certificate of accuracy with NIST traceability
  • Ideal for calibration of measuring equipment or for direct gaging of parts
  • Constructed from durable tool steel
  • Features a 4 micro coating/finish for enhanced performance