Learn More
MSC Vermont Gage Pin Gage, 0.1135 in, 2 in, Class X Minus
Catalog No. 5000527473
Encompass
This Class X Minus Pin Gage has a diameter of 0.1135 in and a length of 2 in. It features a 4 Micro finish and is constructed from tool steel.
  • For calibration of measuring equipment or to gage parts directly.
  • Includes certificate of accuracy with NIST traceability.
  • Made from tool steel for durability.
  • Features a 4 Micro finish.
Catalog No. 50-005-27473 Supplier MSC Supplier No. 88811351
This Class X Minus Pin Gage has a diameter of 0.1135 in and a length of 2 in. It features a 4 Micro finish and is constructed from tool steel.
  • For calibration of measuring equipment or to gage parts directly.
  • Includes certificate of accuracy with NIST traceability.
  • Made from tool steel for durability.
  • Features a 4 Micro finish.