Learn More
MSC Vermont Gage Pin Gage, 0.118 in, Class ZZ Go Plus, Tool Steel
Catalog No. 5000529654
Encompass
Vermont Gage Black Guard Class ZZ Plus Pin Gages are designed for precise measurement and extended lifespan. They feature a black oxide treatment that penetrates the surface, preventing rust and corrosion while providing visible wear indication for effective calibration or replacement.
  • Class ZZ Go (Plus) tolerance
  • NIST traceable
  • Constructed from tool steel with 60/65 Rc hardness
  • Features a 10 microinch finish or better
  • Round within 0.0001 in
  • Certificate of accuracy included
  • Marked with size and direction of tolerance
  • Individually packaged
  • Prevents rust and corrosion
  • Offers visible wear indication for calibration
  • Provides longer product life
Catalog No. 50-005-29654 Supplier MSC Supplier No. 03529831
Vermont Gage Black Guard Class ZZ Plus Pin Gages are designed for precise measurement and extended lifespan. They feature a black oxide treatment that penetrates the surface, preventing rust and corrosion while providing visible wear indication for effective calibration or replacement.
  • Class ZZ Go (Plus) tolerance
  • NIST traceable
  • Constructed from tool steel with 60/65 Rc hardness
  • Features a 10 microinch finish or better
  • Round within 0.0001 in
  • Certificate of accuracy included
  • Marked with size and direction of tolerance
  • Individually packaged
  • Prevents rust and corrosion
  • Offers visible wear indication for calibration
  • Provides longer product life